Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172258.3 | NP_758462.2 | 9 | 1232 | ||
GenBank mRNA | AF453745.1 | - | 9 | 1232 | ||
AK132910.1 | - | 10 | 1318 | |||
AY211261.1 | - | 9 | 1214 |