Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153139.4 | NP_694779.3 | 11 | 1567 | ||
GenBank mRNA | AF453743.1 | - | 10 | 1553 | ||
AY211262.1 | - | 11 | 1537 | |||
BC138556.1 | - | 11 | 1531 | 8861576 | ||
BC138558.1 | - | 11 | 1531 | 8861578 |