Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35e3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029875.2 | NP_084151.2 | 5 | 972 | ||
GenBank mRNA | AK145213.1 | - | 5 | 465 | ||
BC006601.1 | - | 5 | 991 | 3965585 | ||
BC057101.1 | - | 5 | 972 | 5720169 |