Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cnksr3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172546.2 | NP_766134.1 | 6, 7 | 1228 | ||
GenBank mRNA | AK033015.1 | - | 6, 7 | 1229 | ||
AK149951.1 | - | 5, 6 | 527 | |||
AY151137.1 | - | 6, 7 | 1190 | |||
BC024086.1 | - | 5, 6 | 576 |