Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arfgef3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033258.4 | NP_001028430.1 | 22 | 3814 | ||
GenBank mRNA | AK122475.1 | - | 11 | 2176 | ||
AK147679.1 | - | 22 | 3815 | |||
AK147963.1 | - | 22 | 3812 | |||
AK148343.1 | - | 22 | 3759 | |||
AK165602.1 | - | 22 | 3756 |