Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam26f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175449.4 | NP_780658.2 | 1, 2 | 583 | ||
GenBank mRNA | AK042270.1 | - | 1, 2 | 583 | ||
AK080048.1 | - | 1, 2 | 218 | |||
BC095946.1 | - | 1, 2 | 597 | 4945137 |