Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178679.2 | NP_848794.1 | 3 | 1097 | ||
GenBank mRNA | AF543761.1 | - | 2 | 818 | ||
AJ516033.1 | - | 3 | 959 | |||
AK031846.1 | - | 3 | 893 | |||
AK033892.1 | - | 3 | 859 | |||
AK044322.1 | - | 3 | 856 | |||
AK044655.1 | - | 2 | 180 | |||
AK077316.1 | - | 3 | 870 | |||
AK122387.1 | - | 3 | 1084 | |||
BC080272.1 | - | 3 | 1057 | 6837597 | ||
BC157983.1 | - | 3 | 898 | 9088140 | ||
BC157991.1 | - | 3 | 898 | 9088144 |