Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153543.2 | NP_705771.2 | 2 | 225 | ||
GenBank mRNA | AK052361.1 | - | 2 | 116 | ||
AK086413.1 | - | 2 | 167 | |||
AK141954.1 | - | 2 | 187 | |||
BC034531.1 | - | 2 | 147 | 4188055 | ||
CJ130150.1 | - | 2 | 225 |