Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153543.2 | NP_705771.2 | 5 | 738 | ||
GenBank mRNA | AK052361.1 | - | 5 | 629 | ||
AK086413.1 | - | 5 | 681 | |||
AK141954.1 | - | 5 | 700 | |||
AK215056.1 | - | 2 | 51 | |||
BC034531.1 | - | 5 | 660 | 4188055 |