Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172553.4 | NP_766141.1 | 4 | 1050 | ||
GenBank mRNA | AK047846.1 | - | 4 | 1051 | ||
AK081538.1 | - | 3 | 475 | |||
AK086731.1 | - | 4 | 863 | |||
BC052200.1 | - | 4 | 1032 | 6513129 |