Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab3ip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003950.2 | NP_001003950.1 | 2 | 323 | ||
GenBank mRNA | AK152122.1 | - | 2 | 375 | ||
BC080289.1 | - | 2 | 323 | 6816077 |