Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Os9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171026.1 | NP_001164497.1 | 6 | 663 | ||
NM_177614.3 | NP_808282.2 | 6 | 663 | |||
GenBank mRNA | BC006844.1 | - | 6 | 483 | ||
BC031768.1 | - | 6 | 628 | 4487829 | ||
BF470029.1 | - | 1 | 31 | |||
BU703218.1 | - | 6 | 627 |
Gene Symbol: Gm16491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014574.1 | - | 1 | 2547 |