Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Os9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171026.1 | NP_001164497.1 | 10 | 1132 | ||
NM_177614.3 | NP_808282.2 | 10 | 1132 | |||
GenBank mRNA | AK028047.1 | - | 10 | 1107 | ||
BC006844.1 | - | 10 | 952 | |||
BC031768.1 | - | 10 | 1097 | 4487829 | ||
CX566458.1 | - | 4 | 251 |
Gene Symbol: Gm16491 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014574.1 | - | 1 | 1162 |