Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003913.2 | NP_001003913.1 | 15 | 2016 | ||
NM_001171582.1 | NP_001165053.1 | 15 | 2016 | |||
GenBank mRNA | AK190213.1 | - | 1 | 62 | ||
AK191034.1 | - | 1 | 62 | |||
AK198349.1 | - | 1 | 62 | |||
AK201611.1 | - | 1 | 62 | |||
BC052042.1 | - | 15 | 1868 | |||
BC058968.1 | - | 15 | 1908 | |||
BC079643.1 | - | 15 | 1869 | 6416029 |