Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcte1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013688.2 | NP_038716.2 | 4 | 1467 | ||
GenBank mRNA | BC145741.1 | - | 3 | 1274 | 40131044 | |
M28821.1 | - | 3 | 1274 |