Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aftph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290545.1 | NP_001277474.1 | 9 | 2889 | ||
NM_181411.4 | NP_852076.1 | 8 | 2808 | |||
GenBank mRNA | AK030572.1 | - | 8 | 2752 | ||
AK047225.1 | - | 7 | 1872 | |||
AK136815.1 | - | 7 | 3623 | |||
AK150458.1 | - | 6 | 599 | |||
AK152308.1 | - | 6 | 599 | |||
AK152487.1 | - | 6 | 599 | |||
BC004630.1 | - | 7 | 1097 | |||
BC039995.1 | - | 3 | 266 | |||
BC052036.1 | - | 8 | 2736 | 5702534 |