Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aftph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252503.2 | NP_001239432.1 | 2 | 1733 | ||
NM_001290545.1 | NP_001277474.1 | 2 | 1733 | |||
NM_181411.4 | NP_852076.1 | 2 | 1733 | |||
GenBank mRNA | AK029681.1 | - | 2 | 1632 | ||
AK030572.1 | - | 2 | 1677 | |||
AK047225.1 | - | 1 | 797 | |||
AK136815.1 | - | 1 | 985 | |||
AK138793.1 | - | 2 | 1655 | |||
BC004630.1 | - | 1 | 22 | |||
BC052036.1 | - | 2 | 1661 | 5702534 |