Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papolg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172555.2 | NP_766143.2 | 17, 18 | 1911 | ||
GenBank mRNA | AK035810.1 | - | 17, 18 | 1894 | ||
BC019635.1 | - | 2, 3 | 264 | |||
BC023030.1 | - | 2, 3 | 248 | |||
BC037439.1 | - | 2, 3 | 296 | |||
BC049163.1 | - | 17, 18 | 1857 | |||
BC059271.1 | - | 17, 18 | 1905 | 2811338 | ||
DQ103505.1 | - | 17, 18 | 1717 |