Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc85a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166661.2 | NP_001160133.1 | 3 | 2042 | ||
NM_181577.5 | NP_853555.2 | 3 | 2042 | |||
GenBank mRNA | AK045568.1 | - | 3 | 2015 | ||
AK173298.1 | - | 3 | 2059 | |||
BC020949.1 | - | 2 | 1005 | |||
BC066065.1 | - | 4 | 1481 | 6850852 |