Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc85a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166661.2 | NP_001160133.1 | 1, 2 | 1036 | ||
NM_181577.5 | NP_853555.2 | 1, 2 | 1036 | |||
GenBank mRNA | AK045568.1 | - | 1, 2 | 1009 | ||
AK087049.1 | - | 1, 2 | 979 | |||
AK087070.1 | - | 1, 2 | 979 | |||
AK173298.1 | - | 1, 2 | 1053 | |||
BC066065.1 | - | 1, 2 | 559 | 6850852 |