Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mfap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145426.2 | NP_663401.2 | 3 | 1122 | ||
NM_180599.1 | NP_850930.1 | 3 | 1086 | |||
GenBank mRNA | AK042988.1 | - | 3 | 1070 | ||
AK076071.1 | - | 3 | 1088 | |||
AK155122.1 | - | 3 | 1100 | |||
BC006828.1 | - | 3 | 1081 | 3597878 | ||
BC058567.1 | - | 1 | 2209 |