Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175001.3 | NP_778166.2 | 5 | 333 | ||
GenBank mRNA | AK077615.1 | - | 5 | 327 | ||
AK086913.1 | - | 5 | 312 | |||
AK164417.1 | - | 5 | 312 | |||
BC061191.1 | - | 5 | 323 | 6703228 | ||
BY790081.1 | - | 5 | 334 |