Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175001.3 | NP_778166.2 | 3, 4 | 237 | ||
GenBank mRNA | AK077615.1 | - | 3, 4 | 231 | ||
AK086913.1 | - | 3, 4 | 216 | |||
AK164417.1 | - | 3, 4 | 216 | |||
BC061191.1 | - | 3, 4 | 227 | 6703228 | ||
BY790081.1 | - | 3, 4 | 238 |