Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tom1l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039092.3 | NP_001034181.1 | 9 | 1040 | ||
NM_001039093.1 | NP_001034182.1 | 9 | 1040 | |||
NM_153080.2 | NP_694720.2 | 9 | 1040 | |||
GenBank mRNA | AF467887.1 | - | 9 | 1020 | ||
AK043095.1 | - | 9 | 1020 | |||
AK054531.1 | - | 9 | 1008 | |||
AK166040.1 | - | 9 | 1040 | |||
AK179324.1 | - | 1 | 44 | |||
BC062947.1 | - | 9 | 1004 | 30536491 | ||
BC127266.1 | - | 9 | 1006 | 30357295 |