Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004143.4 | NP_001004143.2 | 6, 7 | 1009 | ||
GenBank mRNA | AK034492.1 | - | 6, 7 | 984 | ||
AK084022.1 | - | 2, 3 | 167 | |||
AK155071.1 | - | 6, 7 | 1010 | |||
AK160992.1 | - | 6, 7 | 918 | |||
AK220368.1 | - | 6, 7 | 970 | |||
BC052540.1 | - | 2, 3 | 304 | |||
BC080737.1 | - | 6, 7 | 880 | 30431735 |