Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099288.1 | NP_001092758.1 | 13, 14 | 1450 | ||
NM_175003.3 | NP_778168.2 | 13, 14 | 1450 | |||
GenBank mRNA | AK079081.1 | - | 8, 9 | 751 | ||
BC056366.1 | - | 13, 14 | 1450 | 6825221 | ||
BC059911.1 | - | 13, 14 | 1450 | 6825221 |