Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntrob | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172560.3 | NP_766148.2 | 10 | 2222 | ||
GenBank mRNA | AK036710.1 | - | 9 | 2042 | ||
AK147774.1 | - | 10 | 2108 | |||
BC058512.1 | - | 10 | 1055 | 6817014 |