Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlgn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198862.2 | NP_942562.2 | 4 | 1179 | ||
GenBank mRNA | AK136883.1 | - | 5 | 1745 | ||
AK173159.1 | - | 4 | 1179 | |||
BE952199.1 | - | 4 | 383 |