Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017376.3 | NP_059072.1 | 2 | 583 | ||
NM_153484.3 | NP_705617.2 | 2 | 490 | |||
GenBank mRNA | AF194420.1 | - | 2 | 517 | ||
AK042146.1 | - | 2 | 744 | |||
AK045575.1 | - | 2 | 439 | |||
AK053229.1 | - | 2 | 428 | |||
AK075601.1 | - | 1 | 141 | |||
AK141606.1 | - | 2 | 411 | |||
AK145125.1 | - | 2 | 552 | |||
AK147965.1 | - | 2 | 516 | |||
AK154472.1 | - | 2 | 528 | |||
AK177827.1 | - | 2 | 294 | |||
AK181094.1 | - | 2 | 295 | |||
AK196541.1 | - | 1 | 114 | |||
AK200544.1 | - | 1 | 115 | |||
AK214482.1 | - | 2 | 294 | |||
AK219791.1 | - | 1 | 109 | |||
AY540631.1 | - | 2 | 569 | |||
AY540632.1 | - | 2 | 384 | |||
BC017689.1 | - | 2 | 346 | 4242534 | ||
BC036982.1 | - | 2 | 547 | 4976241 |