Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017376.3 | NP_059072.1 | 2 | 528 | ||
NM_153484.3 | NP_705617.2 | 2 | 435 | |||
GenBank mRNA | AF194420.1 | - | 2 | 462 | ||
AK042146.1 | - | 2 | 689 | |||
AK045575.1 | - | 2 | 384 | |||
AK053229.1 | - | 2 | 373 | |||
AK075601.1 | - | 1 | 86 | |||
AK141606.1 | - | 2 | 356 | |||
AK145125.1 | - | 2 | 497 | |||
AK147965.1 | - | 2 | 461 | |||
AK154472.1 | - | 2 | 473 | |||
AK177827.1 | - | 2 | 239 | |||
AK181094.1 | - | 2 | 239 | |||
AK185529.1 | - | 2 | 239 | |||
AK196541.1 | - | 1 | 59 | |||
AK200544.1 | - | 1 | 60 | |||
AK214482.1 | - | 2 | 239 | |||
AK219791.1 | - | 1 | 54 | |||
AY540631.1 | - | 2 | 514 | |||
AY540632.1 | - | 2 | 329 | |||
BC017689.1 | - | 2 | 291 | 4242534 | ||
BC036982.1 | - | 2 | 492 | 4976241 |