Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190376.1 | NP_001177305.1 | 14 | 2545 | ||
NM_001190378.1 | NP_001177307.1 | 14 | 2545 | |||
NM_001190379.1 | NP_001177308.1 | 13 | 2473 | |||
NM_178116.4 | NP_835217.1 | 14 | 2560 | |||
GenBank mRNA | AK039670.1 | - | 14 | 2372 | ||
AK122400.1 | - | 12 | 2201 | |||
AK150186.1 | - | 14 | 2398 | |||
AK169118.1 | - | 13 | 2312 | |||
BC023962.1 | - | 9 | 2359 | |||
BC027039.1 | - | 3 | 269 | |||
BC027385.1 | - | 6 | 1351 | |||
BC049133.1 | - | 14 | 2389 | 6827800 | ||
BC056395.1 | - | 14 | 2374 | 6852349 | ||
BC059065.1 | - | 13 | 2340 |