Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276383.1 | NP_001263312.1 | 13 | 1450 | ||
NM_153060.3 | NP_694700.2 | 13 | 1798 | |||
GenBank mRNA | AK156007.1 | - | 14 | 1444 | ||
BC011467.1 | - | 13 | 1798 | |||
BC025823.1 | - | 3 | 910 |