Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tekt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282006.1 | NP_001268935.1 | 5 | 685 | ||
NM_001282007.1 | NP_001268936.1 | 5 | 634 | |||
NM_011569.3 | NP_035699.2 | 5 | 637 | |||
GenBank mRNA | AF081947.1 | - | 5 | 628 | ||
AK005801.1 | - | 5 | 686 | |||
AK076858.1 | - | 5 | 616 | |||
BC050967.1 | - | 5 | 635 | |||
BC098181.1 | - | 5 | 595 | 6312556 |