Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Proca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045516.2 | NP_001038981.2 | 4 | 1062 | ||
GenBank mRNA | AK160258.1 | - | 4 | 1063 |