Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172562.3 | NP_766150.1 | 16 | 1375 | ||
GenBank mRNA | AK083467.1 | - | 5 | 406 | ||
AK135802.1 | - | 2 | 144 | |||
AK157938.1 | - | 16 | 1368 | |||
BC005610.1 | - | 13 | 1101 | |||
BC025448.1 | - | 11 | 867 | |||
BC038821.1 | - | 16 | 1378 | 3990023 |