Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kat7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195003.1 | NP_001181932.1 | 13 | 1772 | ||
NM_001195004.1 | NP_001181933.1 | 12 | 1682 | |||
NM_177619.3 | NP_808287.1 | 11 | 1505 | |||
GenBank mRNA | AK147968.1 | - | 12 | 1499 | ||
AK170389.1 | - | 12 | 1629 | |||
BC024092.1 | - | 11 | 1345 | |||
BC048904.1 | - | 11 | 1460 | 6492124 | ||
BC057102.1 | - | 12 | 1610 | 6401353 | ||
DQ076247.1 | - | 13 | 1700 |