Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081220.1 | NP_001074689.1 | 11 | 5781 | ||
GenBank mRNA | BC058657.1 | - | 1 | 823 |