Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145434.4 | NP_663409.2 | 5 | 1524 | ||
GenBank mRNA | AK041047.1 | - | 5 | 1380 | ||
AK053029.1 | - | 1 | 1545 | |||
AK135640.1 | - | 5 | 1386 | |||
AK137398.1 | - | 5 | 1411 | |||
AK137582.1 | - | 5 | 1389 | |||
AK146430.1 | - | 5 | 1481 | |||
AK154931.1 | - | 5 | 1525 | |||
AK155597.1 | - | 5 | 1525 | |||
BC008989.1 | - | 5 | 969 | 3585478 |