Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172565.2 | NP_766153.1 | 2 | 1553 | ||
GenBank mRNA | AK029101.1 | - | 2 | 1554 | ||
AK169910.1 | - | 3 | 1450 | |||
BC011167.1 | - | 1 | 477 | |||
BC111873.1 | - | 2 | 1549 | 40059437 |