Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195088.1 | NP_001182017.1 | 9 | 1353 | ||
NM_001195089.1 | NP_001182018.1 | 9 | 1252 | |||
NM_001195090.1 | NP_001182019.1 | 9 | 1249 | |||
NM_181856.2 | NP_862904.1 | 9 | 1249 | |||
GenBank mRNA | AK143862.1 | - | 9 | 1357 | ||
AK156764.1 | - | 9 | 1210 | |||
AK171772.1 | - | 9 | 1235 | |||
AK220211.1 | - | 6 | 721 | |||
AY236501.1 | - | 9 | 1161 | |||
AY263160.1 | - | 8 | 1137 | |||
BC138254.1 | - | 8 | 1125 | 8861273 | ||
BC145210.1 | - | 8 | 1128 | 9053761 |