Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153082.4 | NP_694722.2 | 4 | 580 | ||
GenBank mRNA | AK049300.1 | - | 4 | 526 | ||
AY035893.1 | - | 4 | 367 | |||
BC042643.1 | - | 4 | 495 | 5357595 | ||
BC051140.1 | - | 4 | 542 | 6595101 | ||
CB247441.1 | - | 3 | 294 |