Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trappc12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161410.1 | NP_001154882.1 | 2 | 444 | ||
NM_001161411.1 | NP_001154883.1 | 2 | 670 | |||
NM_178811.4 | NP_848926.3 | 2 | 670 | |||
GenBank mRNA | AK031763.1 | - | 2 | 594 | ||
AK052245.1 | - | 2 | 480 | |||
AK079067.1 | - | 2 | 464 | |||
AK084171.1 | - | 2 | 516 | |||
AK086217.1 | - | 2 | 600 | |||
AK132166.1 | - | 2 | 456 | |||
AK147985.1 | - | 2 | 444 | |||
AK171540.1 | - | 2 | 480 | |||
AK182249.1 | - | 1 | 104 | |||
BC024077.1 | - | 2 | 613 | 5097580 | ||
BC030887.1 | - | 2 | 438 | 4483611 |