Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178681.4 | NP_848796.2 | 11 | 1346 | ||
GenBank mRNA | AK032509.1 | - | 11 | 1505 | ||
AK038544.1 | - | 11 | 1074 | |||
AK044516.1 | - | 11 | 1156 | |||
AK047682.1 | - | 12 | 1282 | |||
AK083222.1 | - | 12 | 1357 | |||
AK122355.1 | - | 10 | 834 | |||
AK139408.1 | - | 11 | 1347 | |||
BC070461.1 | - | 11 | 1178 | 30649561 |