Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mbip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145442.2 | NP_663417.1 | 6 | 837 | ||
GenBank mRNA | AK077125.1 | - | 6 | 758 | ||
AK154318.1 | - | 6 | 766 | |||
AK171164.1 | - | 6 | 766 | |||
BC002277.1 | - | 6 | 823 | 3496688 |