Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: L2hgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145443.2 | NP_663418.1 | 8 | 1095 | ||
GenBank mRNA | AK145228.1 | - | 8 | 1094 | ||
AK152450.1 | - | 8 | 1072 | |||
AK168429.1 | - | 8 | 1043 | |||
BC016226.1 | - | 8 | 1039 | 4487011 |