Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009358.3 | NP_033384.2 | 8 | 1070 | ||
GenBank mRNA | AB053464.1 | - | 8 | 1015 | ||
AF316616.2 | - | 8 | 927 | |||
AK154050.1 | - | 8 | 1046 | |||
AK160587.1 | - | 8 | 1011 | |||
AK180636.1 | - | 1 | 102 | |||
AY338234.1 | - | 8 | 1000 | |||
BC010716.1 | - | 8 | 983 | 2655861 |