Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cirh1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011574.2 | NP_035704.2 | 7 | 858 | ||
GenBank mRNA | AK144455.1 | - | 7 | 843 | ||
AK146263.1 | - | 7 | 834 | |||
AK150862.1 | - | 7 | 833 | |||
AK168470.1 | - | 7 | 808 | |||
AK173318.1 | - | 6 | 799 | |||
AK179441.1 | - | 2 | 90 | |||
AK187526.1 | - | 3 | 307 | |||
AK195627.1 | - | 3 | 312 | |||
BC027399.1 | - | 3 | 357 | 4948207 | ||
BC068113.1 | - | 7 | 847 | 5707865 |