Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cipc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289429.1 | NP_001276358.1 | 4 | 651 | ||
NM_001289430.1 | NP_001276359.1 | 3 | 550 | |||
NM_001289431.1 | NP_001276360.1 | 4 | 724 | |||
NM_001289432.1 | NP_001276361.1 | 4 | 598 | |||
NM_173735.3 | NP_776096.2 | 3 | 497 | |||
GenBank mRNA | AK122546.1 | - | 3 | 426 | ||
AK148415.1 | - | 2 | 427 | |||
AK154476.1 | - | 3 | 438 | |||
AK160481.1 | - | 4 | 651 | |||
AK160643.1 | - | 3 | 423 | |||
BC026384.1 | - | 3 | 429 | 4168224 | ||
CF530959.1 | - | 3 | 496 | |||
CN526809.1 | - | 3 | 445 |