Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem63c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172583.2 | NP_766171.1 | 12, 13 | 1103 | ||
GenBank mRNA | AK030062.1 | - | 12, 13 | 1095 | ||
AK034410.1 | - | 11, 12 | 1026 | |||
AK043536.1 | - | 11, 12 | 1024 | |||
BC056936.1 | - | 12, 13 | 1103 | 5695989 | ||
BC057088.1 | - | 12, 13 | 1103 | 5695989 |