Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081170.1 | NP_001074639.1 | 8, 9 | 848 | ||
NM_001291444.1 | NP_001278373.1 | 8, 9 | 848 | |||
NM_001291445.1 | NP_001278374.1 | 7, 8 | 758 | |||
GenBank mRNA | AK030003.1 | - | 3, 4 | 216 | ||
AK036581.1 | - | 7, 8 | 734 | |||
AK054000.1 | - | 8, 9 | 823 | |||
AK122326.1 | - | 6, 7 | 537 |